Umbellulone ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
(-)-Umbellulone
AG-CN2-0085-M010
102987-290EA
193.85
USD
102987-290
Umbellulone ≥98% (by HPLC)
Umbellulone
Offensive principle of the so-called "headache tree". Causes a painful cold sensation. Selective TRPA1 activator. Weak TRPM8 activator. Releases CGRP (calcitonin gene-related peptide) after TRPA1 activation which leads to activation of trigeminovascular pathways and CGRP-mediated neurogenic vasodilatation relevant to produce headache/migraine.
Formula:
C₁₀H₁₄O MW: 150.2 g/mol Storage Temperature: Freezer |
CAS Number:
546-78-1 |
Specification Test Results
Source/Host | Isolated from Umbellularia californica Nutt. |
Purity |
|
Appearance/Color | Colourless oil. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...