Pseudolaric acid B ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: Pseudolarix acid B

AG-CN2-0083-M001 AG-CN2-0083-C100
102987-282EA 34.62 USD
102987-282 102987-284
Pseudolaric acid B ≥98% (by HPLC)
Pseudolaric acid B

Antifungal and antifertility compound. Antitumor compound. PPARalpha signaling agonist. Angiogenesis inhibitor. Apoptosis and autophagy inducer. Microtubule-destabilizing agent. Anti-inflammatory. Inhibits NF-kappaB and p38 signaling. Immunosuppressive.

Formula: C₂₃H₂₈O₈
MW: 432.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01746346
CAS Number: 82508-31-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Isolated from Pseudolarix kaempferi.
Purity 98% (HPLC)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO, ethanol, methanol or chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR