Fostriecin sodium salt ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
(6R)-5,6-Dihydro-6-[(1E,3R,4R,6R,7Z,9Z,11E)-3,6,13-trihydroxy-3-methyl-4-(phosphonooxy)-1,7,9,11-tridecatetraenyl]-2H-pyran-2-one sodium salt
, Fostriecin
Antibiotic
Source/Host: Isolated from Streptomyces pulveraceous subsp. fostreus.
Purity: >98% (HPLC)
Appearance/Color: Colorless solid.
Solubility: Soluble in water, methanol or ethanol.
Anticancer compound. Antifungal. Catalytic inhibitor of topoisomerase II (IC50 = 40 µM). Potent protein phosphatase 2A (PP2A) (IC50 = 1.5 nM) and 4 (PP4) (IC50 = 3 nM) inhibitor. Weak protein phosphatase type 1 (PP1) inhibitor (IC50=131µM). No apparent inhibition on PP2B. Mitotic entry checkpoint inhibitor. Mediates cell cycle arrest at G2-M-phase. Ischemia protective. The PP2A binding site is different from that of okadaic acid
Formula:
C₁₉H₂₆O₉PNa Storage Temperature: Freezer |
CAS Number:
87860-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...