Sclerotiorin ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS

AG-CN2-0054-MC05 AG-CN2-0054-M005 AG-CN2-0054-M001
102987-864EA 131.54 USD
102987-864 102987-866 102987-868
Sclerotiorin ≥97% (by HPLC)
Sclerotiorin

Antibiotic


Source/Host: Isolated from fungus Penicillium sp.
Purity: >97% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in DMSO, ethanol or DMF. Sparingly soluble in aqueous buffers.


Antibacterial. Weak antagonist of human endothelin receptor A (ETA) and B (ETB). Cholesteryl ester transfer protein (CETP) activity inhibitor. Grb2-Shc binding inhibitor. Potent aldose reductase inhibitor. Potent reversible, non-competitive soybean lipoxygenase-1 (LOX-1) inhibitor. Antioxidant. Free radical scavenger. Antifungal.

Storage Temperature: Freezer
CAS Number: 549-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR