LysoGlow84™ ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: 4-(Naphthalene-2-yl)-1H-imidazo[4,5-c]pyridine . TFA

AG-CMA-1005-M001
102981-162EA 366.92 USD
102981-162
LysoGlow84™ ≥98% (by NMR)
LysoGlow84™

Cell/membrane permeable, non-toxic, pH-sensitive fluorescent dye (blue-green range) for live imaging of pH alteration in acidic organelles, lysosomes, autophagolysosomes, vesicles, cells, tissue and even small, transparent whole animals without side effects. Useful as intracellular pH indicator with a wide range of fluorescence (pH 3 to pH 13). pH-dependent dual-emission peaks, can be used for ratiometric evaluation of pH (F400/440). No esterases are involved, therefore no toxic by-products occur. Long-term stable; is barely metabolized. Fluorescence co-localizes with LysoTracker® Red DND-99. (LysoTracker® is a registered trademark of Molecular Probes, Inc.)

Formula: C₁₆H₁₂N₃·C₂F₃O₂
MW: 359.3 g/mol
Storage Temperature: Freezer
CAS Number: 1782983-52-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color Light yellow solid.
Solubility Soluble in DMSO, methanol or other polar solvents. Poorly soluble in water at pH 8 to pH 12.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR