Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Variable Speed Cutting Mill, Eberbach

Supplier: EBERBACH CORP
E3503
Pick one of the strongest cutting mills on the market! Multiple cutting blade options allow for a wide variety of milling options. Everything you need to mill your product is included with purchase.

  • High torque
  • 5" dia. chamber
  • Digital controller
  • 5 year warranty

The E3503 Mid-Size Benchtop Cutting Mill features a 5 inch diameter sealed cutting chamber with four stationary blades (optional six stationary bladed cutting chamber is available) and a rotating cutting head with four replaceable rotating cutting blades that revolve at high speed to produce a shearing action which minimizes heat build-up (optional cutting chamber temp senors are available).

Samples introduced through the hopper are swept around by the rotor until they are cut to sufficient fineness to pass through sieve. The hard, tool steel cutting edges on the knives allow milling of a wider range of materials, including plastics, animal feed, bones, cables, cardboard, electronic scrap, feed pellets, foils, food, leather, material mixtures, non-ferrous metals, paper, pharmaceutical products, refuse derived fuels, rubber, waste, wood and so much more.

The E3503 is designed with a machine tool type of cartridge spindle, small powerful servo motor which allows for variable speed operation of 0 - 3,000 rpm at 115V and 0 - 5,000 rpm at 230V with high torque. The chamber door comes standard with a door safety switch. The machine comes standard with mounting locations for bolting directly to a benchtop or an optional table top stand or floor stand can be purchased.

Accessories information: Note: The stand pictured is not included (Model E3500.A.ST).

Ordering information: Model E3503 is supplied with the following; One stainless steel hopper including cover and wooden plunger, one rotating cutting head with one set of rotating cutting blades, one set of stationary blades, three (3) stainless steel sieves (0.5 mm, 1 mm and 2 mm), one jar adapter, one 16oz. collection jar, and cleaning brush.



 

ORDER

Expand All / Collapse All
Description Speed Supplier No. VWR Catalog Number Unit Price Quantity
Cutting Mill 115V: 0–3000 rpm; 203V: 0–5000 rpm E3503 75836-416 Each Retrieving
Product is energy and/or water efficient
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Sample Volume (nominal volume) 5 - 250 g
Environmentally Preferable Attribute Product is energy and/or water efficient
Dimensions 25 x 20 x 17" (Without Stand)
Weight 254 lbs.
Electrical 115/230V, 50/60Hz
Shipping Weight 275 lbs.
Torque 10.6 Nm (Peak); 4.89 Nm (Continuous)
Chamber Dimensions 5" Dia
Shipping Dimensions 30 x 30 x 20"
Sample input 1.4" Dia.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty