Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9kHz - 2GHz Spectrum Analyzer

Supplier: SAELIG COMPANY INC SE
SSA3021X
An ideal choice for R&D, education, production, maintenance, and other RF applications.

  • 10 Hz 3dB resolution bandwidth
  • Low phase noise
  • Very sensitive with a low noise floor, DANL
  • Excellent amplitude accuracy
  • Wide frequency range
  • Low third-order intercept point
  • Large display

Spectrum Analyzer covers signals in the 9 kHz – 2.1 GHz range and measures the magnitude of an input signal versus frequency over the full frequency range of the instrument. It is ideal for measuring the power of the spectrum of both known and unknown signals. Analyzing electrical signal spectra helps characterize wireless transmitters or other electronic devices, determining dominant frequency, power, distortion, harmonics, bandwidth, and other spectral components of a signal that are not easily detected in time domain displays. The Spectrum Analyzer is light weight (10lb) and small size (15.5" x 8.2" x 4.6"), with a user friendly interface on a large 10.1" TFT LCD. It features 256MB of internal storage memory, with up to 32GB supported externally via the USB port.

See the Data manual for more detailed specifications.

Ordering information: Includes Quick Start, Power Cord, USB Cable, CD (Including Quick Start, Data Sheet and Application Software), Calibration Certificate.



 

ORDER

Expand All / Collapse All
Description Input Voltage Supplier No. VWR Catalog Number Unit Price Quantity
9kHz - 2GHz Spectrum Analyzer 110V/240V, AC SSA3021X 470227-432 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty