You Searched For: VTX-27
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: VTX-27 is a potent and selective inhibitor of the PKCtheta enzyme, with over 1000-fold against classical isoforms and over 10000-fold against atypical...