You Searched For: GNE-477
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: GNE 477 is a potent and efficacious dual PI3K/mTOR inhibitor with an IC(50) value of 4nM and 21nM respectively.