You Searched For: NIBR-17
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: NIBR-17 is a pan class I PI3K inhibitor with IC(50) values of 1, 9.2, 9 and 20nM, respectively for PI3Kalpha - delta.