You Searched For: VX-745
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 5-(2,6-Dichlorophenyl)-2-((2,4-difluorophenyl)thio)-6H-pyrimido[1,6-b]pyridazin-6-one, Purity: 98%, CAS Number: 209410-46-8, Appearance: Form: powder, Storage: Sealed in dry, Store in freezer, under -20 deg C, Size: 5mg