You Searched For: Akt-I-1
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: The compound (Akt-I-1) inhibits only Akt1 (IC(50) 4.6 µM) and does not inhibit AKT2, or AKT3. The compound is a reversible inhibitor, and exhibits a l...