You Searched For: 7-Diethylamino-3-[N-(4-maleimidoethyl)carbamoyl]coumarin
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Thiol-reactive fluorescent probe for protein labeling. MDCC has been used for preparing specific probes for inorganic phosphate.