    Sort by:
  • Search Within Results
You Searched for: "5-dtaf-hydrochloride"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   5-DTAF is highly reactive with proteins. Unlike other reactive fluoresceins, 5-DTAF not only reacts with amino groups, but also reacts with thiol grou...