You Searched For: DAF-2T
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Reference material for DAF-2 (CDX-D0084). Spectral data: lambdaex 495nm; lambdaem 515nm.