You Searched For: 7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin (CPM)
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Widely used blue fluorescent thiol-reactive dye (Ex/Em: 384/470nm). This maleimide derivative of coumarin is essentially non-fluorescent until it reac...