    Sort by:
  • Search Within Results
You Searched for: "5-cfda-n-succinimidyl-ester"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   5-CFDA is membrane-permeant and can be loaded into cells via incubation. Once inside the cells, 5-CFDA is hydrolyzed by intracellular esterases to 5-c...