    Sort by:
  • Search Within Results
3-Carboxy-6,8-difluoro-7-hydroxycoumarin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   This amine-reactive reagent can be used to create blue-fluorescent bioconjugates with excitation/emission maxima ~410/455 nm that are excitable by the...