    Sort by:
  • Search Within Results
N-(3-Bromopropyl)-7-nitro-2,1,3-benzoxadiazol-4-amine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Building block for synthesis. Used for the preparation of fluorescent probes.