    Sort by:
  • Search Within Results
You Searched for: "mpac-br"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   HPLC-precolumn-derivatization reagent for carboxylic acids. Compared to classical BMC it emits at longer wavelength and has stronger fluorescence.