    Sort by:
  • Search Within Results
5-(Azidomethyl)-1,3-dihydroxybenzene
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Building block for synthesis. Applied for the construction of dendrimers containing 1,2,3 triazole rings as connectors via click chemistry with a trip...