You Searched For: AMCA-H-N-succinimidyl ester
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: AMCA-H NHS ester reacts with primary amine groups on antibodies, proteins, peptides and other biomolecules at pH 7.0-9.0. Suitable for labeling lysyl ...