    Sort by:
  • Search Within Results
5-Methylmellein
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Widespread fungal pentaketide. Weak antibacterial, antifungal and antiviral activity. Specific inhibitor of human DNA polymerase lambda. Weak antigerm...