You Searched For: Rubrofusarin
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Monomer of aurofusarin. Mycotoxin. Anticancer and antimycobacterial compound in vivo. Moderate tyrosinase inhibitor. Anti-estrogenic.