    Sort by:
Aphidicolin-17-acetate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Phytotoxin (inhibits root growth of seeding). Specific inhibitor of eukaryotic DNA polymerase alpha and DNA synthesis (competition with dCTP).