    Sort by:
IWR-1-endo
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Potent and reversible cell permeable Wnt pathway signaling inhibitor. Inhibits Wnt-induced accumulation of beta-catenin, leading to proteasomal degrad...