You Searched For: 22-NHC hydrochloride
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Aza-steroid. Hedgehog signaling inhibitor. Competes with 20(S)-OHC for access to the cysteine-rich domain in Smoothened (Smo). For control compound se...