    Sort by:
  • Search Within Results
You Searched for: "adiporon-hydrochloride-hydrate"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   Orally-active adiponectin receptor (AdipoR) agonist. Binds to AdipoR1 and AdipoR2 at low µm concentration. Activates 5'-adenosine monophosphate-activa...