    Sort by:
  • Search Within Results
You Searched for: "theacrine"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   Purine alkaloid related to caffeine. Potent antioxidant. Not through direct scavenging of ROS but strengthen antioxidant systems in vivo. Anti-inflamm...