    Sort by:
  • Search Within Results
You Searched for: "cyclosporin-c"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   Immunosuppressant. Calcineurin inhibitor with lower activity than Cyclosporin A (AG-CN2-0079). Antifungal. Antitumor.