You Searched For: Calpinactam
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Specific anti-mycobacterial. Inhibits growth of Mycobacteria smegmatis but also Mycobacteria tuberculosis. M. smegmatis is used as a model for mycobac...