    Sort by:
  • Search Within Results
Dihydrochlamydocin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Phytotoxin. Derivative of HDAC inhibitor chlamydocin Similar chemical structure to the TAN-1746 compounds (potent HDAC inhibitors).