    Sort by:
  • Search Within Results
You Searched for: "rubratoxin-a"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Adipogen
Description:   Mycotoxin. Potent and selective competitive protein phosphatase 2A (PP2A) inhibitor. Does not inhibit PP1, PP2B, PTP1B, ALP and CIP. Antitumor agent. ...