You Searched For: Umbellulone
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (102987-290)
Supplier: Adipogen
Description: Offensive principle of the so-called "headache tree". Causes a painful cold sensation. Selective TRPA1 activator. Weak TRPM8 activator. Releases CGRP ...