You Searched For: Thiocolchicine
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Antimitotic alkaloid. Tubulin polymerization and microtubule assembly inhibitor. Axonal cytoskeleton modulator. Inhibitor of autophagosome-lysosome fu...