    Sort by:
  • Search Within Results
LysoGlow84™
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (102981-162)
Supplier:  Adipogen
Description:   Cell/membrane permeable, non-toxic, pH-sensitive fluorescent dye (blue-green range) for live imaging of pH alteration in acidic organelles, lysosomes,...