You Searched For: n-Dodecyl-alpha-D-maltoside
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Adipogen
Description: Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enz...