    Sort by:
  • Search Within Results
Ingenol-3,20-dibenzoate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89161-416)
Supplier:  Enzo Life Sciences
Description:   Ingenol is a diterpenoid related to phorbol. Activates protein kinase C (PKC) and competes with [3H]phorbol dibutyrate for binding to PKCα (Ki for bin...