    Sort by:
  • Search Within Results
5(S)-Hydroxyeicosa-6E,8Z,11Z,14Z-tetraenoic acid (HETE)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Biosynthesis via 5-lipoxygenase. Chemotactic and chemokinetic for PMNL. Induces degranulation and calcium flux.