You Searched For: 5(S),12(R)-Dihydroxyeicosa-6E,8E,10E,14Z-tetraenoic acid
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (89157-394)
Supplier: Enzo Life Sciences
Description: Biosynthesis via non enzymatic and enzymatic hydrolysis of LTA₄ and by myeloperoxidase-dependent metabolism of cysteinyl leukotrienes. Neutrophil chem...