    Sort by:
  • Search Within Results
R-(+)-IAA-94
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89152-394)
Supplier:  Enzo Life Sciences
Description:   Potent mammalian skeletal muscle and epithelial chloride channel blocker (Ki=1 μM in bovine kidney cortex microsomes).