    Sort by:
  • Search Within Results
8-(4-Chlorophenylthio)-2'-O-methyladenosine-3’,5’-cyclic monophosphate sodium salt
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Specific, potent and membrane permeant activator of the Epac cAMP receptor.