    Sort by:
  • Search Within Results
PTMIO, free radical
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89151-236)
Supplier:  Enzo Life Sciences
Description:   Free radical similar to 4-amino-TEMPO and 3-carboxy-2,2,5,5-tetramethyl-1-pyrrolidine-1-oxyl, but more lipophilic than the former compounds.