You Searched For: Phalloidin
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (76481-780)
Supplier: AAT BIOQUEST INC
Description: Phalloidin, a bicyclic heptapeptide toxin, binds specifically at the interface between F-actin subunits, locking adjacent subunits together.
Minority or Woman-Owned Business Enterprise Small Business Enterprise