    Sort by:
  • Search Within Results
Silver two part conductive adhesive, EXP. XX/XX resin and hardener
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite Expiration date provided