    Sort by:
Silicon surface sensitizer, gold chloride 0.5%
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Applications: Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.