    Sort by:
  • Search Within Results
Perfluoro(methyldecahydronaphthalene)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Trifluoromethyl(perfluorodecahydronaphthalene). Grade:tech. 85, Melting Point C-70*. Boiling Point C:159-160*. C11F20. 51294-16-7. Balance other fluorocarbons.