    Sort by:
  • Search Within Results
Palladium ruthenium on alumina
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Uses: Reduction of nitrogen oxides to N2 with H2 used in presence of CO and CO2