    Sort by:
  • Search Within Results
4-Phenylazomaleinanil
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   4-(N-Maleimido)azobenzene. Grade: 95, Melting Point C162-165*. Boiling Point C: NA. C16H11N3O2. 16201-96-0. IRRITANT