    Sort by:
  • Search Within Results
4-Methyl-3-nitrophenyl isocyanate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (AAAL12669-06)
Supplier:  Alfa Aesar
Description:   Grade: 96. Melting Point C51-53*. Boiling Point C: 113-118*/3mm. C8H6N2O3. 13471-69-7. HARMFUL IRRITANT SENSITISING KEEP COLD MOISTURE SENSITIVE